This item is now discontinued. Please browse similar items
Minntech 279-20-019 Hollow fiber cartridge filter; 0.1 µm, 20"L
Representative image only.
Mfr # 279-20-019 – Item # WZ-29980-06
Provide high flow rates with low pressure drop
  • Steam sterilize capsules in-line or autoclave off-line
  • Polysulfone membrane offers low protein binding
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolysulfone
  • Pore Size (micron)0.1
  • Filter Size508
  • Housing MaterialPolycarbonate
  • Connectionsreplacement filter only
  • Typical Water Flow Rate22.8 LPM
  • Max Pressure (PSI)30
  • Max Pressure2
  • DescriptionHollow fiber cartridge filter; 0.1 µm, 20\"L
  • Warranty1 year
1 Year Warranty
Key Features
  • Steam sterilize capsules in-line or autoclave off-line
  • Polysulfone membrane offers low protein binding
  • 1 year warranty
More About this Item
These new hollow fiber filters offer superior performance in a compact size. Filters have up to six times more surface area compared to other pleated membrane filters, providing greater capacity and longer filter life. The unique membrane structure helps remove bacteria, high-level endotoxins, pyrogens, and spores — ideal for a variety of water purification applications. Maximum differential pressure is 30 psi (2 bar). Capsules and cartridges provided nonsterile.