This item is now discontinued. Please browse similar items
Miltex Dissecting-Grade Iris Tissue Forceps; 1/Pk
Representative image only.
Miltex Mfr # V96-120 – Item # WZ-10918-24
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • GradeDissecting
  • Length (in)4
  • Length (cm)10.16
  • End TypeCurved
  • MaterialStainless Steel
  • DescriptionDissecting-Grade Iris Tissue Forceps; 1/Pk
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Dissection-grade instruments are manufactured from stainless steel; hardened, tempered, and polished for a high-priced finish without the added cost. These instruments are well-suited for less critical applications with tight budgets such as necropsies.