This item is now discontinued. We recommend the following as suggested replacements: WZ-10100-23

Mettler Toledo XPR2U Ultra-Micro Balance; 2.1 g

Representative image only.
Mettler Toledo Mfr # XPR2U – Item # WZ-01030-14
Automatically records all measurements via built-in notepad
  • LevelControl warns when the balance is not leveled and proposes corrective action
  • Draft shield dismantles completely for easy cleaning
  • Motorized automatic draft-shield door offers safe access to weighing pan
  • Color capacitive touch screen allows quick “swipe” through of settings and methods
  • Active Temperature Control system increases weighing stability by removing excess heat
  • IR proximity sensors provide programmable touch-free balance control and door operation
  • Multiple ports offer secure connectivity and data export
  • 2.1 g capacity
  • 0.0001 mg readability
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (g)2.1
  • Readability (mg)0.0001
  • Repeatability (mg)0.00025
  • Linearity (mg)0.0015
  • Weighing Unitsg, mg, µg, ct, oz, ozt, GN, dwt, mom, tlh, tls, tlt
  • Calibration TypeInternal
  • LanguagesEnglish, Chinese, Czech, French, German, Hungarian, Italian, Japanese, Polish, Russian, Spanish,
  • OutputUSB (4), LAN
  • USB InterfaceYes
  • Stability IndicatorYes
  • Power (VAC)100 to 240
  • Height (in)7 3/32
  • Width (in)4 27/32
  • Depth (in)14 1/64
  • Pan Diameter (in)5/8
  • DescriptionUltra-Micro Balance, 2.1 g
  • Warranty1 Year
1 Year WarrantyCanadian Standards Association

Includes

keypad, 15-ft (4.5-m) keypad cable, weighing pan, weighing chamber plate, drip tray, glass draft shield, tweezers, cleaning brush, pen, USB storage device, and power cable.

Key Features

  • LevelControl warns when the balance is not leveled and proposes corrective action
  • Draft shield dismantles completely for easy cleaning
  • Motorized automatic draft-shield door offers safe access to weighing pan
  • Color capacitive touch screen allows quick “swipe” through of settings and methods
  • Active Temperature Control system increases weighing stability by removing excess heat
  • IR proximity sensors provide programmable touch-free balance control and door operation
  • Multiple ports offer secure connectivity and data export
  • 2.1 g capacity
  • 0.0001 mg readability
  • 1 year warranty

More About this Item

The compact, space-saving XPR micro balances are ideal for weighing toxic and hazardous substances in a protected environment where user safety demands are high. Programmable infrared (IR) sensors enable touch-free operation of various functions, greatly reducing the risk of cross-contamination. A simple wave of the hand automatically opens the draft-shield door. Another safety feature is the Active Temperature Control (ATC™) system which removes heat from the balances’ electronics resulting in increased weighing stability.

To simplify the handling of samples the balances offer two keypads from which to carry out your weighing tasks. The SmartView keypad located above the weighing chamber offers all the basic weighing functions right at your fingertips. The main keypad which is connected to the balance by a cable, can be placed wherever it is most easily accessible. This remote keypad features a color capacitive touch screen that allows you to swipe through settings, applications, and methods even while wearing thick gloves.

A built-in results notepad automatically records all measurements and comments, eliminating manual transcription. To ensure consistent processes, regularly used task methods are stored in the method library where they can be quickly accessed. Results and task parameters can easily be transferred to a computer or software application via direct transfer or USB flash drive.

The balances are also equipped with tolerance profiles which ensure weighing tasks meet defined quality requirements and regulations. In addition, the GWP® Approved quality assurance function actively monitors the balance to confirm it is safe to weigh—a green light illuminates when the balance is ready to accept samples.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat