Mettler Toledo Ultra Micro-ISM pH Electrode; MultiPin™
Representative image only.
Mettler Toledo Mfr # 30244732 – Item # WZ-35901-40
Expertly-engineered probes to cover a wide range of applications
  • ISM® - Intelligent Sensor Management⌡Cable not included⌡ARGENTHAL™ reference system⌡LoT membrane for low temperature and ionic strength samples
PRODUCT OPTIONS
£498.97GBP / Each
£598.76GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSmall volumes and containers
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteFRISCOLYT-B®
  • Sensing Glass TypeLoT - Low temperature and low ionic strength applications
  • Min pH (pH)1
  • Max pH (pH)11
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ATC Element
  • ConnectorMultipin™
  • Diameter (mm)3
  • Length (mm)40
  • DescriptionpH Electrode; MultiPin™
Key Features
  • ISM® - Intelligent Sensor Management⌡Cable not included⌡ARGENTHAL™ reference system⌡LoT membrane for low temperature and ionic strength samples
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
ISM technology stores calibration data in the sensor and automatically transfer it to the meter.⌡Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors. ⌡Special low-temperature glass membrane (LoT) is ideal for working with low temperature and low ionic strength samples. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.