This item is now discontinued. Please browse similar items

Mettler Toledo SG98 Handheld pH / Ion / RDO Meter Only

Representative image only.
Mettler Toledo Mfr # 51302661 – Item # WZ-35903-80
Rugged, portable, and ergonomic
  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty
Include Accessories Today!
Mettler Toledo Water Analysis Accessories
£9.12 - £446.91GBP / Each
£10.94 - £536.29GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Min pH (pH)-2
  • Max pH (pH)19.999
  • pH Resolution0.001
  • pH Accuracy±0.002
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1
  • mV Accuracy±0.1
  • Min ISE0
  • Max ISE9999 ppm
  • ISE Resolution3 or 4 digits
  • ISE Accuracy±0.5%
  • Min Dissolved Oxygen0
  • Max Dissolved Oxygen50 ppm
  • Dissolved Oxygen Resolution0.01
  • Dissolved Oxygen Accuracy±0.1 mg/L up to 8 mg/L, ±0.2 mg/L from 8 to 20 mg/L, ±10% above 20 mg/L
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.2°C
  • Temperature compensation-5 to 130°C (ATC), -30 to 130°C (MTC)
  • OutputUSB, RS-232, Infrared
  • Data Logging (points)500 measurements (GLP)
  • WaterproofYes
  • BatteryFour 1.5 V AA alkaline batteries
  • Length (cm)22
  • Width (cm)9
  • Height (cm)4.5
  • Width (in)3 1/2
  • Length (in)8 11/16
  • Height (in)1 3/4
  • DescriptionHandheld pH / Ion / RDO Meter Only
  • Warranty1 year
1 Year Warranty

Key Features

  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty

More About this Item

The SevenGo Duo Pro handheld multiparameter meter displays both measured parameters simultaneously. All measurements are stored with time-and-date stamp, electrode and sample data, and other relevant information to ensure GLP guidelines are met. Download saved data to PC via RS-232 or USB (order adapter cables separately).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat