This item is now discontinued. Please browse similar items

Mettler Toledo SG78 Handheld pH / Ion / Conductivity Meter Field Kit

Representative image only.
Mettler Toledo Mfr # 51302622 – Item # WZ-35903-78
Rugged, portable, and ergonomic
  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty
Include Buffers Today!
Oakton® pH Buffer Solution Packs
£34.25 - £370.14GBP / Pkg of 3
£41.10 - £444.17GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Min pH (pH)-2
  • Max pH (pH)19.999
  • pH Resolution0.001
  • pH Accuracy±0.002
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1
  • mV Accuracy±0.1
  • Min ISE0
  • Max ISE9999 ppm
  • ISE Resolution3 or 4 digits
  • ISE Accuracy±0.5%
  • Min Conductivity0.01 µS/cm
  • Max Conductivity1000 mS/cm
  • Conductivity Resolution0.01
  • Conductivity Accuracy±0.5% of reading
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.2°C
  • Temperature compensation-5 to 130°C (ATC), -30 to 130°C (MTC)
  • OutputUSB, RS-232, Infrared
  • Data Logging (points)500 measurements (GLP)
  • WaterproofYes
  • Battery4 AA (included)
  • Length (cm)22
  • Width (cm)9
  • Height (cm)4.5
  • Width (in)3 1/2
  • Length (in)8 11/16
  • Height (in)1 3/4
  • DescriptionHandheld pH / Ion / Conductivity Meter Field Kit
  • Warranty1 year
1 Year Warranty

Includes

InLab® Expert Pro-ISM pH electrode and InLab 738-ISM conductivity cell, both with IP67 and 6-ft (1.8-m) cables, uGo™ carrying case, wrist strap, 4 AA batteries, and test certificate supplied by the manufacturer.

Key Features

  • Ergonomic design provides easy right-or left-handed operation
  • IP67-rated housing is watertight and dustproof, including sensor connections
  • Automatic end point, sensor, and buffer recognition functions simplify operation
  • 1 year warranty

More About this Item

The SevenGo Duo Pro handheld multiparameter meter displays both measured parameters simultaneously. All measurements are stored with time-and-date stamp, electrode and sample data, and other relevant information to ensure GLP guidelines are met. Download saved data to PC via RS-232 or USB (order adapter cables separately).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat