Mettler Toledo D5 Excellence Density Meter, 100-240 VAC
Representative image only.
Mettler Toledo Mfr # D5 – Item # WZ-11335-29
Accurate, intuitive, and modular with the One Click user interface
  • Powerful autosamplers for all sample types, including high-viscosity solutions
  • Simultaneous determination of density, refractive index, pH, conductivity and color
  • LabX PC software complies with 21 CFR Part 11
  • Accuracy of ±0.00005 g/cm3
  • 1 year warranty
PRODUCT OPTIONS
£19,502.48GBP / Each
£23,402.98GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Width (InchShort)15 5/32
  • Height (in)15 5/32
  • Density Resolution (g/cm³)0.00001 g/cm3
  • Density Accuracy±0.00005 g/cm3
  • Width (cm)43.87
  • Width (in)15 5/32
  • DescriptionDensity meter Excellence D5
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Includes
Density meter, external power supply, country-specific power cable, terminal WVGA7 inch AnaChem, syringe adapter, waste tube, three disposable syringes, combined water standard (9 mL), user manual, declaration of conformity, and test report.
Key Features
  • Powerful autosamplers for all sample types, including high-viscosity solutions
  • Simultaneous determination of density, refractive index, pH, conductivity and color
  • LabX PC software complies with 21 CFR Part 11
  • Accuracy of ±0.00005 g/cm3
  • 1 year warranty
More About this Item
Excellence Density Meters guarantee high reliability and maximum precision, with an accuracy of up to 6 digits. Measure density, specific gravity, as well as many related values such as % alcohol and Brix quickly. The touch screen terminal with intuitive One Click user interface minimizes training and operator influence. Ensure precise density measurements with the integrated video view and BubbleCheck™. Automatic viscosity correction combined with integrated temperature control ensures trustworthy results for all samples. The density meter and refractometer can be stacked, enabling you to combine a density meter with a refractive index cell to measure both parameters at once.