This item is now discontinued. Please browse similar items
Mettler Toledo 738-ISM-10M Conductivity Cell, standard range, IP67, ISM; 10 m cable
Representative image only.
Mettler Toledo Mfr # 51344114 – Item # WZ-35902-48
Outstanding performance for any application
  • Integrated temperature compensation sensor ensures measurements are accurate
  • Choose from a variety of materials and cable lengths to suit your requirements
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 0.57
  • Cell TypeContact
  • Electrode MaterialGraphite
  • Body MaterialEpoxy
  • Min temperature (° C)0
  • Max temperature (° C)100
  • Min temperature (° F)32
  • Max temperature (° F)212
  • Cable Length (m)10
  • Cable Length (ft)33
  • DescriptionConductivity Cell, standard range, IP67, ISM; 10 m cable
Key Features
  • Integrated temperature compensation sensor ensures measurements are accurate
  • Choose from a variety of materials and cable lengths to suit your requirements
More About this Item
These laboratory conductivity cells are all dip style with shaft materials ranging from plastic to metal to glass, and graphite, stainless steel, or platinum electrode bands. Common shaft diameter of 12 mm is compatible with most electrode holders. Rugged IP67-rated cells are preferred for outdoor and industrial use.
Intelligent Sensor Management (ISM) conductivity cells feature onboard electronic memory that stores calibration data, unique identification number, and temperature history. When connected to your compatible conductivity meter, the sensor is automatically recognized and the stored data uploads to the meter for immediate use, saving time and preventing errors.