This item is now discontinued. We recommend the following as suggested replacements: WZ-01018-60
Mettler Toledo 211214 Filter Pan Suitable For 50 mm Filters
Representative image only.
Mettler Toledo Mfr # 211214 – Item # WZ-11140-29
Keep your balance clean and your samples contained
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)1 31/32
  • DescriptionFilter Pan Suitable For 50 mm Filters
More About this Item
This kit features a special weighing pan that allows precise positioning of the filters. The glass fiber filters are the only ones officially supported by Mettler Toledo moisture analyzers for optimal use and guaranteed performance.