Mettler Toledo 11107765 Point Electrode For Discharging Small Samples For XS Analytical Balances
Representative image only.
Mettler Toledo Mfr # 11107765 – Item # WZ-11140-94
  • Remove electrostatic charge from samples and tare containers
PRODUCT OPTIONS
£1,032.97GBP / Each
£1,239.56GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeAntistatic Probe
  • DescriptionPoint Electrode For Discharging Small Samples For XS Analytical Balances
Key Features
  • Remove electrostatic charge from samples and tare containers
More About this Item
Find pans, hooks, temperature kits, and more for your balances and misture analyzers.