This item is now discontinued. Please browse similar items
Meta 6116-19-SZ Unisex Poly-Cotton Lab Coat, Lt Blue, X-Large (46-48)
Representative image only.
Meta Mfr # 6116-19-SZ XL – Item # WZ-10505-06
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)40
  • Length (cm)101.6
  • MaterialPolyester / Cotton
  • SizeX-Large
  • ColorLight Blue
  • DescriptionUnisex Poly-Cotton Lab Coat, Lt Blue, X-Large (46-48)
More About this Item
Coats are made of stain- and odor-resistant 65% polyester, 35% cotton poplin. Side-slits allow easy access to pants pockets. Coats are 40" long and feature three spacious pockets and an adjustable back belt.