This item is now discontinued. We recommend the following as suggested replacements: WZ-86468-40
Meta 17010-011-S Labcoat, Fluid Resistant, Ladies, S
Representative image only.
Meta Mfr # 17010-011-S – Item # WZ-86468-01
Liquid quickly beads and rolls off fabric, helping to keep lab coats dry and stain-free!
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)40
  • Length (cm)101.6
  • MaterialPolyester / Cotton
  • SizeSmall
  • ColorWhite
  • DescriptionLabcoat, Fluid Resistant, Ladies, S
More About this Item
Lab coats are constructed of 65% polyester/35% cotton fabric and treated with NANO-TEX™ protection at a molecular level. Molecular structures, known as nano-whiskers, are attached to the tiny fabric fibers—permanently altering the performance of the fabric. This allows the fabric to repel liquids and stains without altering the look, feel, or comfort of the lab coat. Ideal for biotech, pharmaceutical, and general laboratory use.
Coats feature a five-button front and one interior (lower right) pocket to maximize functionality. One-piece back includes a two-button swing belt for added style and a better fit. Coats also feature three exterior pockets and one interior (lower right) pocket to maximize functionality. White collar.