This item is now discontinued. Please browse similar items
Meriam ZA900529-00014 HART Lead Kit with Standard Assembly
Representative image only.
Meriam Mfr # ZA900529-00014 – Item # WZ-23001-40
Optimize performance of your calibrator
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHART Lead Kit with Standard Assembly
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Choose from a wide range of accessories to enhance operation of your Meriam calibrator.
Kit is included with MFC and is composed of standard banana plugs, mini-grabbers, & 250 ohm resistor.