This item is now discontinued. Please browse similar items
Meriam 30EBX25-020-01-2-02 Ruggedized Well-Type Table-Mount Manometers, Scale A, 20" WC
Representative image only.
Meriam Mfr # 30EBX25-020-01-2-02 – Item # WZ-68460-12
Accurate and dependable, as well as highly versatile
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% of smallest graduation
  • Min Pressure (in WC)0
  • Max Pressure (in WC)20
  • Liquid FillWater
  • Process Connection1/4\" NPT(F)
  • Media CompatibilityAir and noncorrosive gases
  • DescriptionRuggedized Well-Type Table-Mount Manometers, Scale A, 20\" WC
  • Warranty1 year
1 Year Warranty
Key Features
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
  • 1 year warranty
More About this Item
Well-type manometers provide fast, direct readings of pressure, vacuum, differential, flow and liquid levels. No adding of distances above and below a zero line. All scales are graduated
to correct for the drop in well level and are full width of the instrument.
Ranges: 6, 12, 20, 30, 40, and 60 inches