This item is now discontinued. Please browse similar items
Memosens CA/MS-010NAA Cables, 10 M ( Digital Sensors for Knick Meters)
Representative image only.
Memosens Mfr # CA/MS-010NAA – Item # WZ-56030-36
Plug-and-play digital sensor technology for pH, conductivity and dissolved oxygen
  • Add additional parameters by simply adding optional Memosens sensors
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Cables
  • DescriptionCables, 10 M ( Digital Sensors for Knick Meters)
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Add additional parameters by simply adding optional Memosens sensors
  • 1 year warranty
More About this Item
The Memosens sensors digitally store their serial number, electrode type, unique ID, and calibration data. Easily change your meter from DO to pH to conductivity with Memosens smart sensor recognition. The sensors can also be moved from one meter to another and transfer their calibration data.