Meiji Techno Microscope Camera Adaptor, C-Mount, 0.3X, for 1/3" Chip Camera
Representative image only.
Meiji Techno Mfr # MA151/35/03 – Item # WZ-48400-63
Easily connect your Meiji Techno microscope to a camera
  • Compatible with Meiji Techno microscopes
PRODUCT OPTIONS
£285.81GBP / Each
£342.97GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Eyepiece Magnification0.3
  • SizeFits over 21 mm reticle
  • DescriptionMicroscope Camera Adaptor, C-Mount, 0.3X, for 1/3\" Chip Camera
Key Features
  • Compatible with Meiji Techno microscopes
More About this Item
Use C-mount adapters to attach a digital camera or video camera to the photo tube of your Meiji Techno microscope. The C-mount adapter that is needed will depend on your microscope, as well as your camera. Camera and microscope sold separately.