Masterflex Variable-Area In-Line Dual Media/Scale Flowmeters with Valve, Direct-Reading, PTFE Fitting; 0.8 GPM Air/Water
Representative image only.
Masterflex Item # WZ-32053-20
Interchangeable medias and scales offer flexibility
  • Scale rotates to display rates for either air-like or water-like fluids
  • PTFE/glass wetted parts are ideal for aggressive chemicals
  • Breakaway back plate protects staff in case of pressure surge blowout
  • Use for laboratory or research applications requiring high purity
  • Valved units offer both control and indication of flowrate
PRODUCT OPTIONS
£515.73GBP / Each
£618.88GBP (incl VAT)
£281.70GBP / Each
£338.04GBP (incl VAT)
for an additional
£281.70GBP / Each
£338.04GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Repeatability±5% full-scale
  • Max Pressure (PSI)100
  • Max Air Flow Rate (mL/min)100000
  • Max Water Flow Rate (mL/min)3000
  • Max Flow Rate (mL/min)100000
  • Material Flow TubeBorosilicate glass
  • Material FittingPTFE and PCTFE
  • Material ValvePTFE and PCTFE
  • Material O-RingPTFE
  • Material FloatPTFE
  • Material HousingAluminum
  • ValveYes
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • DescriptionVariable-Area In-Line Dual Media/Scale Flowmeters with Valve, Direct-Reading, PTFE Fitting; 0.8 GPM Air/Water
Key Features
  • Scale rotates to display rates for either air-like or water-like fluids
  • PTFE/glass wetted parts are ideal for aggressive chemicals
  • Breakaway back plate protects staff in case of pressure surge blowout
  • Use for laboratory or research applications requiring high purity
  • Valved units offer both control and indication of flowrate
More About this Item
These meters have been designed to measure aggressive water-like liquid solutions and aggressive air-like gase mixtures. The dual water scale shows flow rate in gallons per minute (gpm) and liters per minute (LPM); the dual air scale shows flow rate in standard cubic feet per minute (scfm) and liters per minute (LPM).
Flowmeters are individually tested to 1 x 10-7 sccs of helium or better.