Masterflex Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame; Medium Flow
Representative image only.
Masterflex Item # WZ-03217-88
Control your flow with the addition of a valve
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
PRODUCT OPTIONS
£40.77GBP / Each
£48.92GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Width (")1754
  • Min Air Flow Rate (mL/min)20000
  • Max Air Flow Rate (mL/min)60000
  • Min Water Flow Rate (mL/min)0
  • Max Water Flow Rate (mL/min)1200
  • Max Flow Rate (mL/min)60000
  • Material O-RingBuna-N
  • Material HousingAluminum
  • DescriptionVariable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Medium Flow
Key Features
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
More About this Item
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than mL/min, convert them to mL/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.