Masterflex Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Stainless Steel Fittings without valve, 8.5 L/min Air
Representative image only.
Masterflex Item # WZ-68560-10
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
PRODUCT OPTIONS
£45.09GBP / Each
£54.11GBP (incl VAT)
£59.59 - £281.70GBP / Each
£71.51 - £338.04GBP (incl VAT)
for an additional
£281.70GBP / Each
£338.04GBP (incl VAT)
for an additional
£59.59GBP / Each
£71.51GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Max Pressure (PSI)100
  • Max Flow Rate (LPM)8.5
  • Material Fitting316 SS
  • Material O-RingViton®
  • Material HousingAcrylic
  • ValveNo
  • Wetted Materials316 SS
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Media TypeAir
  • DescriptionAcrylic Flowmeter w/o Valve for Air, SS, 8.5 LPM
Key Features
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
  • Flowmeter features stainless steel fittings and Viton® o-ring
More About this Item
Versatile acrylic flowmeters are designed specifically for air. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.