Masterflex Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 10 mm Tube OD, 24 VDC
Representative image only.
Masterflex Item # WZ-98305-03
Excellent choice for viscous fluids
  • Specifically designed for metric sized tubing
PRODUCT OPTIONS
£148.43GBP / Each
£178.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Pinch Valve Tube Size10 mm OD
  • Connection TypePinch Valve for Tubing
  • Power (VDC)24
  • Watts13
  • Flow PatternTwo-Way, Normally Open
  • DescriptionSolenoid-Operated Two-Way Pinch Valve; Normally-Open, 10 mm Tube OD, 24 VDC
Key Features
  • Specifically designed for metric sized tubing
More About this Item
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept two tubes, the bottom tube is normally open while the top is normally closed.