Masterflex Reusable Water Filtration Element without Housing, 100 µm
Representative image only.
Masterflex Item # WZ-29650-50
Design reduces operating expenses when filtering solids-heavy liquids
  • Easily clean inner core by separating discs and rinsing solids—no tools required
  • Maximum temperature for cartridge without housing is 190°F (88°C)
PRODUCT OPTIONS
£131.25GBP / Each
£157.50GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Micron Rating100
  • Filter Length (in)10
  • DescriptionReusable water filtration element without housing; 100 µm
Key Features
  • Easily clean inner core by separating discs and rinsing solids—no tools required
  • Maximum temperature for cartridge without housing is 190°F (88°C)
More About this Item
The unique design includes a central core holding several hundred precisely grooved polypropylene discs that are tightly stacked together. The stack creates a large internal surface area capable of holding a large volume of solids.
Rinsing the element restores the cartridge to its original condition—saving the cost of cartridge replacement and disposal. Use the 50-μm filters as a prefilter to protect finer disposable filters or carbon cartridges. Use the 100-μm element to remove visible solids. Select the 200-μm element when heavy solids loading is expected.