Masterflex Power Cord, 230 VAC, European Plug; 6-ft Long
Representative image only.
Masterflex Item # WZ-50001-70
Optimize performance of your Masterflex® pump
  • Choose from a wide range of power cords
PRODUCT OPTIONS
£46.70GBP / Each
£56.04GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)230
  • AC Max (Volts)230
  • Description230 VAC power cord with European plug, 6-ft long
Key Features
  • Choose from a wide range of power cords
More About this Item
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.