Masterflex Low Pressure-Drop Flow Switches for Liquids, Closed; 1.0 GPM
Representative image only.
Masterflex Item # WZ-32776-21
Minimize flow constriction and achieve low pressure drop
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 Stainless Steel, and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • Durable switch with 212°F (100°C) max temperature and 200 psi at 70°F max pressure
PRODUCT OPTIONS
£104.01GBP / Each
£124.81GBP (incl VAT)
£301.20GBP / Each
£361.44GBP (incl VAT)
for an additional
£301.20GBP / Each
£361.44GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)212
  • Max Operating Temperature (° C)100
  • DescriptionNormally closed switch for liquids; 1.0 GPM
Key Features
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 Stainless Steel, and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • Durable switch with 212°F (100°C) max temperature and 200 psi at 70°F max pressure
More About this Item
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 µm filter to protect your switch. Switches include 1/4" male quick connect terminals.