Masterflex Flowmeter Tripod Base Stand for 32461-Series Flowmeters
Representative image only.
Masterflex Item # WZ-32462-55
Easily mount flowmeters on tripod for easy viewing
  • Constructed of durable acrylic
  • Suitable for use with 32461-Series flowmeters
PRODUCT OPTIONS
£85.13GBP / Each
£102.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTripod base Flowmeter Stand For 32461
Key Features
  • Constructed of durable acrylic
  • Suitable for use with 32461-Series flowmeters
More About this Item
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.