Masterflex Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, 3/4" Mini x 1/2" NPT(F)
Representative image only.
Masterflex Item # WZ-31200-13
Maintain sanitary conditions
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
Free Certificate of Comliance
£114.07GBP / Each
£136.88GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeSanitary x NPT(F)
  • Thread size NPT(F) (port 2) (in)1/2
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionTubing Adapter; Polypropylene (PP), 3/4" mini to 1/2" NPT (F)
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
  • Connection Type: 3/4" mini to 1/2" NPT(F)
More About this Item
These fittings are ideal for use with the Masterflex L/S tubing pumps.