Masterflex Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
Representative image only.
Masterflex Item # WZ-30600-43
Regulate flow without any leaking
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
£99.70GBP / Pkg of 10
£119.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortFemale Luer
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow Pattern5 Ports, 360 Degree Rotation
  • DescriptionManifolds with luer locks, 5 ports, 360° rotation; 10/pack, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Five ports with female luer locks
More About this Item
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.