Masterflex Fitting, Nylon, Straight, Threaded Plug, 3/4" NPT(M); 10/PK
Representative image only.
Masterflex Item # WZ-30401-46
Deigned for permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
£18.00GBP / Pkg of 10
£21.60GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNylon
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)3/4
  • DescriptionThreaded plug, nylon, 3/4" male NPT, 10/pack
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/4" NPT(M)
More About this Item
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).