Masterflex Fitting, Black PVDF, One-Way, Stopcock with Male Luer Lock to Female Luer Lock; 10/PK
Representative image only.
Masterflex Item # WZ-31200-91
Build leakproof fluid handling systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
£78.30GBP / Pkg of 10
£93.96GBP (incl VAT)
Often Bought with…
Velp MST Digital Magnetic Stirrer; 100-240 VAC, 50-60 Hz
£246.74GBP / Each
£296.09GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsBlack PVDF
  • Flow PatternOne-Way
  • DescriptionLuer stopcock, 1-way, male luer lock x female luer, black PVDF, pack of 10
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: female x male luer lock, one-way flow pattern
More About this Item
These one-way stopcocks feature an all-black PVDF construction.