Masterflex DB9 Male 9-pin Connector; for Creating Your Own Cable
Representative image only.
Masterflex Item # WZ-07595-45
Create your own cable for optimization of your Masterflex® drive
  • Choose the correct connection for your Masterflex® pump
PRODUCT OPTIONS
£76.21GBP / Each
£91.45GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDB9 male 9-pin connector for use with drives.
Key Features
  • Choose the correct connection for your Masterflex® pump
More About this Item
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).