Masterflex DB25 Male Connector; for Creating Your Own Cable
Representative image only.
Masterflex Item # WZ-07523-94
Create your own cable for optimization of your Masterflex® drive
  • Choose the correct connection for your Masterflex® pump
PRODUCT OPTIONS
£92.53GBP / Each
£111.04GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeAnalog Variable Speed
  • DescriptionDB25 male connector; for creating your own cable.
Key Features
  • Choose the correct connection for your Masterflex® pump
More About this Item
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).