This item is now discontinued. Please browse similar items
Master Appliance MT-30 Low profile table-top microtorch with wide, stable base
Representative image only.
Master Appliance Mfr # MT-30 – Item # WZ-03030-52
Low profile, table-top microtorch for industrial and laboratory applications
  • Butane-powered, eliminates the need for external fuel, costly piping or unsightly hoses
  • Meets all consumer products safety commission (CPSC) requirements
  • Adjustable broad flame or pinpoint flame
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • Width (in)3 1/2
  • Length (in)3
  • Height (in)5 7/64
  • DescriptionLow profile table-top microtorch with wide, stable base
  • WarrantyManufacture Defect
Key Features
  • Butane-powered, eliminates the need for external fuel, costly piping or unsightly hoses
  • Meets all consumer products safety commission (CPSC) requirements
  • Adjustable broad flame or pinpoint flame
More About this Item
This refillable table-top microtorch produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). Perfect for shrinking tubing, specialty connectors, spot drying materials, heating wax and many more applications. The torch features a built-in self-igniting system with gas shut-off safety lock, a extra wide base perfect for hands-free use and a durable fuel tank containing enough fuel for up to 60 minutes of usage. Torch measure 5.1" H x 3" W x 3.5" D