This item is now discontinued. Please browse similar items
Master Appliance MT-11K Microtorch Kit with Butane Canister, Up to 2500F, W Ind-Resistant Flame
Representative image only.
Master Appliance Mfr # MT-11K – Item # WZ-03030-50
Compact size is perfect for field service kits
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • DescriptionMicrotorch Kit with Butane Canister, Up to 2500F, W Ind-Resistant Flame
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This refillable microtorch unit produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). The pinpoint flame is perfect for heat shrinking & terminating as well as general heating applications. Unit features a built in safety lock, self-ignition system and meets CPSC safety requirements.
What's included: one Microtorch with 15/16 oz. can of butane