This item is now discontinued. Please browse similar items
Marvel Scientific 6ERF Hazardous Material Undercounter Refrigerator/Freezer, 6 cu ft
Representative image only.
Marvel Scientific Mfr # 6ERF0001 – Item # WZ-44765-75
Refrigerator/Freezers that are appropriate for noncritical temperature storage applications
  • 1 year warranty parts & labor
SPECS & DESCRIPTION
Specifications & Description
  • Refrigerator Capacity (cu ft)6.1
  • Refrigerator Capacity (Liters)173
  • Min Temperature - Refrigerator (° F)35
  • Max Temperature - Refrigerator (° F)42
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)6
  • Number of Doors1
  • Number of Shelves2
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionHazardous Material Undercounter Refrigerator/Freezer, 6 cu ft
  • Warranty1 year parts & labor
1 Year Parts and LaborCanadian Standards AssociationUL Listed.
Key Features
  • 1 year warranty parts & labor
More About this Item
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.
Hazardous Location Models are for use where explosive conditions could potentially exist external to the cold storage unit. These units contain a specially designed compressor with all electrical components enclosed to prevent accidental ignition of refrigerator contents. These products have a UL listing for Class 1, Group C & D applications and must be hard-wired to voltage source.