This item is now discontinued. Please browse similar items
Marsh Bellofram 969-712-000 Pressure Transmitter, I to P;6-30 PSI, 4-20 mA; NEMA4X
Representative image only.
Marsh Bellofram Mfr # 969-712-000 – Item # WZ-68826-22
Install in limited-space locations or manifold
  • 4 to 20 mA Input
  • Small footprint, compact size
  • Built-in volume booster
  • External zero and span adjustments
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Exhaust Flow Rate (SCFM)1
  • Forward Flow Capacity (SCFM)24
  • Forward Flow Capacity (LPM)677
  • Min Operating Temperature (° F)-20
  • Max Operating Temperature (° F)160
  • Min Operating Temperature (° C)-28
  • Max Operating Temperature (° C)71
  • Control Range (low) (PSI)6
  • Control Range (high) (PSI)30
  • Process Connection NPT(F) (in)1/4
  • Electrical Connections½\" conduit
  • ApprovalNEMA 4X
  • DescriptionPressure Transmitter, I to P;6-30 PSI, 4-20 mA; NEMA4X
  • Warranty18 months
18 Month WarrantyCanadian Standards Association
Key Features
  • 4 to 20 mA Input
  • Small footprint, compact size
  • Built-in volume booster
  • External zero and span adjustments
  • 18 month warranty
More About this Item
Transducers are an electro-pneumatic device that reduce supply pressure to a regulated output pressure directly proportional to an electrical input signal. They provide precise control to actuators, valves, and positioners, and are ideally used for high-flow devices. Supply and output ports are located on front and back of units, enabling simple installation and connections. Choose from a general-purpose or NEMA 4X enclosure.