This item is now discontinued. Please browse similar items
Mark-10 TSTME Torque test stand, horizontal, lever-operated, 100 lbFin
Representative image only.
Mark-10 Mfr # TSTME – Item # WZ-59848-03
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)100
  • Capacity (N-m)11.3
  • ResolutionAngle dial with 2° resolution
  • Speed RangeHand operated
  • Depth (in)31 39/64
  • Height (in)9 13/32
  • Width (in)7 29/32
  • DescriptionTorque test stand, horizontal, lever-operated, 100 lbFin
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • 3 year warranty
More About this Item
Manual torque measurement test stands are ideal for a wide variety of torque testing applications up to 100 lbFin, including closure torque testing, fastener torque testing, and more. Multipurpose design allows the stand to be configured for a variety of torque sensors and gripping fixtures. Both test stands feature a lever-operated slider with travel stops to make engaging and disengaging samples quick and efficient. Choose from vertival or horizontal leveral operatred models.
Bottle and cap grips are sold separately for gripping bottles and containers for closure testing.