Mark-10 MR52-50Z Torque Sensor, 50 ozFin / 3600 gFcm / 36 kgFmm / 350 Nmm / 35 Ncm
Representative image only.
Mark-10 Mfr # MR52-50Z – Item # WZ-25302-11
Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
PRODUCT OPTIONS
£941.66GBP / Each
£1,129.99GBP (incl VAT)
£237.42 - £280.70GBP / Each
£284.90 - £336.84GBP (incl VAT)
for an additional
£237.42GBP / Each
£284.90GBP (incl VAT)
for an additional
£280.70GBP / Each
£336.84GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)50
  • Capacity (g-cm)3600
  • Resolution (oz-in)0.02
  • Resolution (g-cm)2
  • Resolution (N-cm)0.02
  • Accuracy±0.35% of full scale + indicator
  • DescriptionPlug & Test Bidirectional Static Torque Sensor; 35 N
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
  • 3 year warranty
More About this Item
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.