This item is now discontinued. Please browse similar items
Mark-10 R51 Plug & Test Closure Torque Sensor; 14 N cm
Representative image only.
Mark-10 Mfr # MR53-20Z – Item # WZ-25302-37
Rugged aluminum housing
  • Reversible jaws for round or rectangular containers
  • Spill proof construction
  • May be used for torque tool calibration
  • Warranty: 3 years
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)20
  • Capacity (g-cm)1400
  • Resolution (oz-in)0.02
  • Resolution (g-cm)1
  • Resolution (N-cm)0.01
  • Accuracy±0.5% of full scale + indicator
  • DescriptionPlug & Test Closure Torque Sensor; 14 N cm
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Reversible jaws for round or rectangular containers
  • Spill proof construction
  • May be used for torque tool calibration
  • Warranty: 3 years
  • 3 year warranty
More About this Item
Ideal for testing torque on bottle caps, closures, valves, fasteners, and other items in sizes ranging between 0.56 - 5.7 [14 - 148 mm] diameter. Models include movable posts to easily accommodate a variety of shapes and sizes.