This item is now discontinued. Please browse similar items
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 7, 1 pair
Representative image only.
MAPA Mfr # 65031-7 – Item # WZ-86357-40
Inner fabric lining provides exceptional comfort
SPECS & DESCRIPTION
Specifications & Description
  • MaterialButyl
  • Thickness (mil)60
  • Size7
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • LiningCotton
  • CuffScalloped
  • SterileNo
  • DescriptionCotton-Lined Butyl Gloves, size 7, 1 pair
More About this Item
Butyl gloves provide excellent chemical resistance in applications involving acids, ketones, esters, and amine derivatives. 100% seamless cotton liner provides extra comfort for long-term use. Scalloped cuffs can be rolled down to prevent drips from reaching your arms. Unique Z-pattern grip design for optimal grip.