This item is now discontinued. View the direct replacement: WZ-70001-02 or browse similar items
MadgeTech TransiTempII-RH Low Cost Temperature and Humidity Data Logger
Representative image only.
MadgeTech Mfr # TRANSITEMPII-RH – Item # WZ-23000-26
TransiTempII, TransiTempII-RH, and Cryo-Temp
  • Programmable alarms and warnings
  • Instantaneous out of range indication
  • IP65 splash proof rated
  • Delayed Start
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeTemperature and humidity
  • Min Temperature (° C)-40
  • Max Temperature (° C)80
  • Temperature Accuracy (° F)±1.0
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.5
  • Humidity Accuracy±5%RH
  • Memory Size16,383/Channel
  • Battery Life1 year - factory replacement
  • Length (in)3 19/64
  • Height (in)2 7/64
  • Width (in)13/32
  • DescriptionLow Cost Temperature and Humidity Recorder
  • Warranty1 year
1 Year WarrantyIP 65
Includes
Lanyard.
Key Features
  • Programmable alarms and warnings
  • Instantaneous out of range indication
  • IP65 splash proof rated
  • Delayed Start
  • 1 year warranty
More About this Item
These splash proof temperature data loggers featuring a highly precise semiconductor temperature sensor and an ABS plastic enclosure. This affordable, N.I.S.T. traceable device is ideal for monitoring and recording the temperature of all types of perishable products, including food items and pharmaceutical goods, both in storage and in transit.
The three available devices are equipped with three status LED indicators to alert you to the temperature status. Green signals safe conditions, yellow indicates warning limits have been exceeded, and red denotes alarm limits have been exceeded. The units can be programmed through software to start immediately, delayed start for up to 30 days, or be set to start using the magnetic switch.
The TransiTempII-RH is a humidity and temperature data logger that is ideal for monitoring and recording the temperature and humidity of products during storage and shipping. The TransiTempII-RH also uses the IFC300 docking station for data download (model 23000-34 sold separately).
No external probes are required to achieve its high performance as both the temperature and humidity sensors are internal to the device. The TransiTempII-RH has an humidity resolution of 0.1% RH and typical accuracy of ±3.5% RH at 25°C. It can record up to 16,383 readings per channel.