This item is now discontinued. Please browse similar items
MadgeTech TEMPRETRIEVER Mini Reusable Temp Logger with Alarm
Representative image only.
MadgeTech Mfr # TEMPRETRIEVER – Item # WZ-18003-25
  • FEATURES
  • Real time operation
  • Low cost
  • User friendldy
  • Battery powered - user replaceable
  • Programmable start time
  • Small size, quick data retrieve
  • Non volatile solid state memory
  • Readings selectable 1 every 2 seconds to 1 every 12 hours
  • Pushbutton start / stop (MH100)
  • 1 year warranty
  • SPECS & DESCRIPTION
    Specifications & Description
    • Number of Channels1
    • Input TypeTemperature
    • Max Temperature (° F)176
    • Min Temperature (° F)-40
    • Min Temperature (° C)-40
    • Max Temperature (° C)80
    • Temperature Accuracy (° C)0.5
    • Memory Size38400
    • Battery Life1 yr
    • Length (in)2 1/5
    • Height (in)1 2/5
    • Depth (in)19/32
    • DescriptionMini Reusable Temp Logger with Alarm
    • Warranty1 year
    1 Year Warranty
    Key Features
    • FEATURES
    • Real time operation
    • Low cost
    • User friendldy
    • Battery powered - user replaceable
    • Programmable start time
    • Small size, quick data retrieve
    • Non volatile solid state memory
    • Readings selectable 1 every 2 seconds to 1 every 12 hours
    • Pushbutton start / stop (MH100)
  • 1 year warranty
  • More About this Item
    Each of these temperature data loggers can be started and stopped directly from a personal computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software allows data to be examined, exported and printed.
    If the temperature goes out of the specified range on the MH2000 unit for more than 10 minutes, the devices red LED indicator will start to flash, signaling an alarm condition. The indicator will continue to flash until the alarm is acknowledged. This feature makes the MH2000 the ideal device for monitoring and verifying that food has been stored at the proper temperature. The device utilizes non-volatile solid-state memory as its storage medium, providing maximum data security even if the battery becomes discharged.
    FEATURES
    • Small size
    • Non-volatile solid state memory
    • Readings selectable 1 every 2 sec to 1 every 12 hours
    • Pushbutton start/stop (MH100 and MH2000)
    • Real-time operation
    • Low cost
    • User-friendly
    • Battery powered - user replaceable
    • Programmable start time