This item is now discontinued. Please browse similar items
M-System L53U-1202-AD4 Multi Power Transducer; 5 A, 4-20mA
Representative image only.
M-System Mfr # L53U-1202-AD4 – Item # WZ-49031-14
Record several parameters simultaneously
  • Measure several variables of a heavy-current power system simultaneously: current, voltage, active, reactive and apparent power, active and reactive energy, power factor, and frequency
  • All measured values, counter values, setting data are stored in the nonvolatile memory at the power off and can be transmitted via RS-485/MODBUS® RTU or analog output
  • Conversion factors, system configuration, and interval times are programmable
  • Single-phase/2-wire and 3-wire, three-phase/3-wire, 4-wire systems
  • Up to 31st harmonic distortion measurement
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)110 to 240
  • DescriptionMulti Power Transducer; 5 A, 4-20mA
  • Warranty3 Years
3 Year Warranty
Key Features
  • Measure several variables of a heavy-current power system simultaneously: current, voltage, active, reactive and apparent power, active and reactive energy, power factor, and frequency
  • All measured values, counter values, setting data are stored in the nonvolatile memory at the power off and can be transmitted via RS-485/MODBUS® RTU or analog output
  • Conversion factors, system configuration, and interval times are programmable
  • Single-phase/2-wire and 3-wire, three-phase/3-wire, 4-wire systems
  • Up to 31st harmonic distortion measurement
  • 3 year warranty
More About this Item
L53U Series Multi Power Transducers
-- Multifunctional power measurement of electric devices or switching boards
-- Program using the PC configuration software
-- DIN rail mounted