This item is now discontinued. Please browse similar items
Lovibond RS5690 Colorimeter Reagent Set, Silica, Low Range, 100 tests
Representative image only.
Lovibond Mfr # 535690 – Item # WZ-99560-87
Premeasured reagents in convenient powder packs
  • Precise dosing with simple push-button operation
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersSilica, low range
  • Number of Tests Included100
  • DescriptionColorimeter Reagent Set, Silica, Low Range, 100 tests
  • Warranty6 month
6 Month WarrantyConformite Europeenne
Key Features
  • Precise dosing with simple push-button operation
  • 6 month warranty
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.