This item is now discontinued. Please browse similar items

Lovibond MD100 Colorimeter, Silica HR Kit

Representative image only.
Lovibond Mfr # 276116 – Item # WZ-99561-96
Uses popular Powder Pack™, tablet, and liquid chemistries
  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeLCD
  • Light SourceLED
  • Absorbance Accuracy±1 nm
  • Test ParametersSilica; high-range
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)40
  • Number of Tests Included100
  • Data LoggingYes
  • Memory Size16 data sets
  • Battery1 x 9V battery (included)
  • Length (in)6 1/8
  • Width (in)3
  • Height (in)1 1/2
  • Range1 to 90 mg/L
  • DescriptionColorimeter, Silica HR Kit
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant
  • 2 year warranty

More About this Item

The Lovibond MD100 series colorimeters are available in a wide variety of parameters using powder-pack chemistries. These rugged, waterproof meters feature a large backlit display, real-time clock, data logging capabilities, and a user calibration mode which allows you to calibrate against prepared standards.

(Note: Lovibond is the new name for Orbeco-Hellige--exactly the same product with a new name.)

Range for reagent: 1 to 90 mg/L

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat