LinkTech 40PP Quick-Disconnect Insert, almond PP, 1/4" flow, In-Line, nonvalved, 1/4" hose barb; 25/pk SKU 4130513
Representative image only.
LinkTech Mfr # 40PP-PE2-04MALD – Item # WZ-41305-13
Easy to disconnect from 1/4" flow line with one hand
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
PRODUCT OPTIONS
£44.62GBP / Pkg of 25
£53.54GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Valve MaterialPolypropylene
  • Valve Spring Material316 SS
  • O-Ring MaterialPeroxide-cured EPDM
  • Sterilization MethodGamma
  • Connection TypeQuick-Disconnect x Barb
  • ColorAlmond
  • Min Temperature (° F)32
  • Max Temperature (° F)160
  • Min Temperature (° C)0
  • Max Temperature (° C)71
  • Max Operating Pressure (PSI)120
  • Fitting Length (mm)32
  • DescriptionPolypropylene Quick-Disconnect Insert, 1/4" Flow, In-Line, Straight-Through 1/4" Hose Barb
Key Features
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
More About this Item
Just press the metal thumb-latch to connect and disconnect fitting halves. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized.
Mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.