This item is now discontinued. Please browse similar items
Lightnin 801074PSP 316 SS axial flow impeller ( A-100), 11.4" dia x 3/4" bore dia
Representative image only.
Lightnin Mfr # 801074PSP – Item # WZ-50350-30
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • Impeller Size (in)11.4
  • Impeller Size (mm)289.56
  • Description316 SS axial flow impeller ( A-100), 11.4\" dia x 3/4\" bore dia
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Axial Flow Impellers (LIGHTNIN® A-100) with SUPERPITCH® design — 1.5 pitch ratio for greater batch mixing capabilities. Standard axial flow; use for wetting dry powders.