Lascar Electronics SGD 24-M420 PanelPilot 2.4" Graphic Panel Meter; 4-20 mA Input
Representative image only.
Lascar Electronics Mfr # SGD 24-M420 – Item # WZ-18000-20
Your fastest route to a personalized display!
  • Display connects to a PC via USB port for customization of color, units, logo, and graphical or numerical representation of measurements
  • Free PanelPilot software download is compatible with Windows® XP, 2000, Vista, and Windows 7
  • Panel mount with the included hardware, or leave free standing
  • Screw terminal connections for scaleable 4 to 20 mA sensor input
  • 1 year warranty
PRODUCT OPTIONS
£126.82GBP / Each
£152.18GBP (incl VAT)
£107.32 - £165.86GBP / Each
£128.78 - £199.03GBP (incl VAT)
for an additional
£107.32GBP / Each
£128.78GBP (incl VAT)
for an additional
£165.86GBP / Each
£199.03GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Control Input mA4-20
  • Display Type2.4\" color touchscreen
  • Max Power VDC (VDC)30
  • Min Power VDC (VDC)4
  • DescriptionPanelPilot 2.4\" Graphic Panel Meter; 4-20 mA Input
  • Warranty1 Year
1 Year Warranty
Key Features
  • Display connects to a PC via USB port for customization of color, units, logo, and graphical or numerical representation of measurements
  • Free PanelPilot software download is compatible with Windows® XP, 2000, Vista, and Windows 7
  • Panel mount with the included hardware, or leave free standing
  • Screw terminal connections for scaleable 4 to 20 mA sensor input
  • 1 year warranty
More About this Item
These versatile color TFT graphic meter displays are user-programmable to show a variety of styles previously supported by traditional panel meters. Easily configure with the free PanelPilot software to display information through bar graph, analog, or numeric meter styles with user-defined color, text, units, and input scaling. A library of predefined elements such as meters, buttons, and switches is constantly being updated and expanded. USB to micro-USB cable is required for PC connection and programming. A standard null-modem cable will not power the unit as well as transfer data for programming, please order the 18000-58 interface cable (not included). Once programmed, the display can be left connected to a PC and powered through the USB port, or disconnected and powered through the 4 to 30 VDC terminal and mounted where needed. A USB to AC power adapter (part number 18000-59) is also available so you may plug the meter into a standard North American wall outlet.
Configuring your PanelPilot display is simple; just follow these five easy steps:
1. Download the PanelPilot software (see zip file in the Technical Resources section)
2. Connect your display to the USB port of your PC using cable 18000-58 (must be purchased separately, please see Accessories section)
3. Customize by selecting from an ever-increasing library of configurations
4. Test your setup on the software before uploading the to your display.
5. Upload the configuration to your meter, and use in your application!