This item is now discontinued. Please browse similar items
Lascar Electronics EL-USB-RT Real-time Temperature & Humidity USB Data Logger
Representative image only.
Lascar Electronics Mfr # EL-USB-RT – Item # WZ-23039-59
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeTemperature and humidity
  • Max Temperature (° F)158
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)±3
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±1.5
  • Max Humidity RH (RH)100
  • Min Humidity RH (RH)0
  • Humidity Resolution (%)1
  • Humidity Accuracy±4.5% RH
  • Memory Size7 days worth
  • Battery LifeUSB Powered
  • Display TypeNo
  • DescriptionReal-time Temperature & Humidity USB Data Logger
  • Warranty1 year
1 Year Warranty
Key Features
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
These data loggers measure temperature and humidity from 31 to 176°F (–35 to 80°C) and 0 to 100% RH, calculate dew point, and store over 16,000 readings per parameter.
Model EL-USB-RT stays permanently connected to your computer USB port—displaying real-time ambient temperature, humidity, and calculated dew point on the PC monitor. Ideal for server room applications, this data logger can be programmed to send an e-mail or text message when a user-selectable alarm has been reached.
Real-time measurements will display on the monitor without creating an enlarging data file—unit will only download data to the computer memory when directed to do so. Logger must be plugged into the computer USB port to acquire readings as it is USB powered; there is no separate battery.
What's included: configuration software and battery.