This item is now discontinued. Please browse similar items
Lascar Electronics EL-USB-5 Event, State Change and Counter Data Logger, Standard Memory
Representative image only.
Lascar Electronics Mfr # EL-USB-5 – Item # WZ-18000-34
Download directly to PC via built-in USB
  • Programmable alarms alert to problems
  • Easily manipulate and analyze data with included software
  • IP67-rated housing protects logger from soil and moisture (cap must be fitted)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeVoltage
  • Max Frequency (Hz)100
  • Min Frequency (Hz)0
  • Sampling Rate0 to 2 Hz (event/state change) or 0 to 100 Hz (count frequency)
  • Memory Size21510
  • InterfaceSoftware
  • ConnectionsUSB
  • Battery Life1 year
  • Battery Type1/2 AA
  • DescriptionEvent, state change & counter data logger
  • Warranty1 year
1 Year Warranty
Key Features
  • Programmable alarms alert to problems
  • Easily manipulate and analyze data with included software
  • IP67-rated housing protects logger from soil and moisture (cap must be fitted)
  • 1 year warranty
More About this Item
—Rising voltage, falling voltage, or volt-free trigger options
—Log up to 2 state changes/second or 4 events/second