Lascar Electronics EL-USB-2-LCD+ High Accuracy Data Logger, RH/Temp/Dew Pt, with LCD Display
Representative image only.
Lascar Electronics Mfr # EL-USB-2-LCD+ – Item # WZ-23039-65
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
  • 1 year warranty
PRODUCT OPTIONS
£145.60GBP / Each
£174.72GBP (incl VAT)
£273.99 - £281.50GBP / Each
£328.79 - £337.80GBP (incl VAT)
for an additional
£281.50GBP / Each
£337.80GBP (incl VAT)
for an additional
£273.99GBP / Each
£328.79GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeTemperature and humidity
  • Max Temperature (° F)176
  • Min Temperature (° F)-31
  • Max Temperature (° C)80
  • Min Temperature (° C)-35
  • Temperature Resolution (° F)1
  • Temperature Accuracy (° F)±0.6
  • Temperature Resolution (° C)0.5
  • Temperature Accuracy (° C)±0.3
  • Max Humidity RH (RH)100
  • Min Humidity RH (RH)0
  • Humidity Resolution (%)0.5
  • Humidity Accuracy±2.0% RH
  • Memory Size16,379
  • Battery Life1 year (typical)
  • Display TypeLCD
  • DescriptionHigh Accuracy Data Logger, RH/Temp/Dew Pt, with LCD Display
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • User-programmable alarm thresholds
  • All data loggers come with software and are great for laboratory environmental monitoring
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
These data loggers measure temperature and humidity from 31 to 176°F (–35 to 80°C) and 0 to 100% RH, calculate dew point, and store over 16,000 readings per parameter.
Model EL-USB-2-LCD+ features a 2.5-digit LCD showing the latest logged values, min/max log, and have immediate, delayed, push-to-start logging, and increased humidity measurement accuracy.
What's included: configuration software and battery.