This item is now discontinued. Please browse similar items
Labconco Xpert TXE 3746716 REQUIRED Base Stand for Xpert TXE with Casters
Representative image only.
Labconco Mfr # 3746716 – Item # WZ-33512-14
Complete your fume hood assembly with these required and optional components
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)24
  • Exterior Height (in)42
  • DescriptionREQUIRED Base Stand for Xpert TXE with Casters
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Required accessories include base stands, work surfaces, and filters. Optional accessories include shelving and holders.
Telescoping Base Stands
Support XPert TXE Equipment
Enclosures. Epoxy-coated 1.75'' (4.4 cm) tubular steel. Adjustable to eight height positions at installation in 1'' (2.5 cm) increments from 27.5'' (70 cm) to 34.5'' (88 cm). Available with fixed feet or 5'' (13 cm) diameter casters. Shipping weight 100 lbs. (45 kg)